missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saccharin Sodium, Dihydrate, Powder, USP, 99-101%, Spectrum™ Chemical
SO200, 6155-57-3, C7H4NNaO3S·2H2O
$531.63 - $44615.03
Chemical Identifiers
CAS | 6155-57-3 |
---|---|
Molecular Formula | C7H8NNaO5S |
Molecular Weight (g/mol) | 241.19 |
InChI Key | NAUHOSQQKJTUAX-UHFFFAOYSA-M |
IUPAC Name | sodium 1,3-dioxo-3H-1λâ¶,2-benzothiazol-1-olate dihydrate |
SMILES | O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18606119
|
Spectrum Chemical Mfg Cor
SO200100GM |
100 g | Poly Bottle |
Each for $531.63
|
|
||||
18606120
|
Spectrum Chemical Mfg Cor
SO200500GM |
500 g | Poly Bottle |
Each for $1,158.40
|
|
||||
18606121
|
Spectrum Chemical Mfg Cor
SO20025KG |
2.5 kg | Poly Pail |
Each for $3,612.34
|
|
||||
18606122
|
Spectrum Chemical Mfg Cor
SO20012KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18606124
|
Spectrum Chemical Mfg Cor
SO20050KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Saccharin Sodium, Dihydrate, Powder, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
6155-57-3 | |
241.19 | |
sodium 1,3-dioxo-3H-1λâ¶,2-benzothiazol-1-olate dihydrate |
C7H8NNaO5S | |
NAUHOSQQKJTUAX-UHFFFAOYSA-M | |
O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 |
Specifications
6155-57-3 | |
Poly Bottle | |
100 g | |
O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 | |
241.19 | |
USP |
1 | |
C7H8NNaO5S | |
NAUHOSQQKJTUAX-UHFFFAOYSA-M | |
sodium 1,3-dioxo-3H-1λâ¶,2-benzothiazol-1-olate dihydrate | |
99 to 101% |