Learn More
Innovatek S. Aureus Rapid Latex Kits
Innovatek S. Aureus Rapid Latex Kits detect the presence of both clumping factor and protein A
Supplier:  Innovatek 419110913
Catalog No. 361045045
Only null left
Explore available promotions
Description
Description
  • Rapid Latex Slide Agglutination Test detects the presence of both clumping factor, and protein A
  • The identification of S. aureus is generally confirmed by the production of coagulase in the microorganism
  • Tube Coagulase Test detects free coagulase, an extracellular enzyme produced by S. aureus
  • Pure culture suspected of being S. aureus should be used
  • Positive reactions usually take 5-20 seconds after testing procedure is complete
Specifications
Specifications
2°C to 8°C
Single-use
100 Tests/Kit
Allow latex reagents to warm to room temperature before use
S. Aureus Latex, S. Aureus Control Latex, Disposable Test Cards
≈3 min.
Product Suggestions
Product Suggestions