missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2-(Anilinomethyl)pyrrolidine 98.0+%, TCI America™
$255.38 - $810.86
Chemical Identifiers
CAS | 64030-44-0 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.263 |
MDL Number | MFCD00015890 |
InChI Key | MCHWKJRTMPIHRA-NSHDSACASA-N |
Synonym | s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline |
PubChem CID | 6950385 |
IUPAC Name | N-[[(2S)-pyrrolidin-2-yl]methyl]aniline |
SMILES | C1CC(NC1)CNC2=CC=CC=C2 |
Chemical Identifiers
64030-44-0 | |
176.263 | |
MCHWKJRTMPIHRA-NSHDSACASA-N | |
6950385 | |
C1CC(NC1)CNC2=CC=CC=C2 |
C11H16N2 | |
MFCD00015890 | |
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline | |
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline |
Specifications
64030-44-0 | |
126°C | |
MFCD00015890 | |
1760 | |
MCHWKJRTMPIHRA-NSHDSACASA-N | |
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline | |
6950385 | |
≥98.0% (T) | |
(S)-(+)-2-(Anilinomethyl)pyrrolidine |
Yellow | |
C11H16N2 | |
1 g | |
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline | |
C1CC(NC1)CNC2=CC=CC=C2 | |
176.263 | |
176.26 | |
Liquid |
Safety and Handling
TSCA : No