missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl Hydrogen Phosphate 97.0+%, TCI America™
$29.83 - $98.79
Chemical Identifiers
CAS | 35193-64-7 |
---|---|
Molecular Formula | C20H13O4P |
Molecular Weight (g/mol) | 348.294 |
MDL Number | MFCD00010045 |
InChI Key | JEHUZVBIUCAMRZ-UHFFFAOYSA-N |
Synonym | 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide |
PubChem CID | 99589 |
SMILES | C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B1144100MG
|
TCI America
B1144100MG |
100 mg |
Each for $29.83
|
|
|||||
B11441G
|
TCI America
B11441G |
1 g |
Each for $98.79
|
|
|||||
Chemical Identifiers
35193-64-7 | |
348.294 | |
JEHUZVBIUCAMRZ-UHFFFAOYSA-N | |
99589 |
C20H13O4P | |
MFCD00010045 | |
1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O |
Specifications
35193-64-7 | |
C20H13O4P | |
100 mg | |
JEHUZVBIUCAMRZ-UHFFFAOYSA-N | |
348.294 | |
348.29 | |
Crystalline Powder |
White-Yellow | |
MFCD00010045 | |
1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O | |
99589 | |
≥97.0% (T) | |
(S)-(+)-1,1′-Binaphthyl-2,2′-diyl Hydrogen Phosphate |
Safety and Handling
TSCA : No