Learn More
RKI Oxygen Sensor for RKI Gas Monitors DFS Item
For use with GX82, GX86, GX-82A, GX-86A and GX-91B portable gas monitors.
Supplier:  RKI 651051RK
Catalog No. 19-023754
Only null left
Explore available promotions
Description
Description
  • Sensors are mounted in the diffusion head at the bottom of the unit
  • The diffusion grill protects the sensors; grill easily snaps off the instrument case so you can conveniently replace the sensors
  • Oxygen (O2) sensor is contained in a cylindrical metal shell, with a bayonet pin base for easy replacement
  • Shell forms one electrical contact, and a button on the base is the other contact
  • The perforations at the end of the shell allow atmosphere to diffuse into the sensor
Specifications
Specifications
GX82, GX86, GX-82A, GX-86A, GX-91B Portable Gas Monitors
Oxygen Sensor
Product Suggestions
Product Suggestions