Learn More
BUCHI Reveleris™ Empty Solid Loader Tube/Frit
Buchi Reveleris™ Empty Solid Loader Tube/Frit is designed for loading a stationary phase and sample to this cartridge for dry loading chromatography applications
Supplier:  BUCHI 145142051
 View more versions of this product
Catalog No. 05-402-912
Only null left
Explore available promotions
Description
Description
  • Loads your stationary phase and sample to this cartridge for dry loading chromatography applications
  • Use supplied frits to hold sample in place
  • Use the proper sized insertion rod (sold separately) to insert frits
Specifications
Specifications
Polypropylene
100/Pk.
Empty Solid Loader
15 g
Product Suggestions
Product Suggestions