Learn More
BUCHI Reveleris™ Empty Solid Loader Tube/Frit
Buchi Reveleris™ Empty Solid Loader Tube/Frit is designed for loading a stationary phase and sample to this cartridge for dry loading chromatography applications
Supplier: BUCHI 145142050
Description
- Loads your stationary phase and sample to this cartridge for dry loading chromatography applications
- Use supplied frits to hold sample in place
- Use the proper sized insertion rod (sold separately) to insert frits
Specifications
Polypropylene | |
100/Pk. | |
Empty Solid Loader | |
3 g |