Learn More
Restek™ Presents SGE Micro-Volume, Positive Displacement Syringes
High precision and accuracy down to 0.1μL.
$377.47 - $462.47
Specifications
Material | PTFE (Tip) |
---|---|
For Use With (Application) | For high pressure injection |
Product Type | SGE Micro-Volume, Positive Displacement Syringes |
Quantity | Ea. |
Catalog Number | Mfr. No. | Needle Gauge | Model | Needle Point Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Gauge | Model | Needle Point Style | Price | Quantity | |||
06712454
![]() |
Restek™
RK24771 |
23 G | 0.5BR-7BV | 2 |
N/A
|
|
|||
06712460
![]() |
Restek™
RK24777 |
23 G | 5BR-5 | Cone |
N/A
|
|
|||
06712458
![]() |
Restek™
RK24775 |
23 G | 1BR-7BV | 2 |
N/A
|
|
|||
06712456
![]() |
Restek™
RK24773 |
23 G | 1BR-5 | Cone |
N/A
|
|
|||
06712462
![]() |
Restek™
RK24779 |
23 G | 5BR-7BV | 2 |
N/A
|
|
|||
Description
- Sample is contained in the needle with full plunger displacement, ensuring no dead volume
- Seal between the plunger and the needle can be tightened for high-pressure injection
- Replaceable needle/plunger kits and barrels
Specifications
PTFE (Tip) | |
For high pressure injection | |
SGE Micro-Volume, Positive Displacement Syringes | |
Ea. |