Learn More
Biochrom Resolution Lite PC Software
Resolution PC Software is an easy to use software platform with powerful modules for measurement and analysis. Resolution can be used with your Libra or WPA spectrophotometer for instrument control increasing your options for working with your spectrophotometer outside of the lab.
Supplier: Biochrom 80710030
Description
- Resolution PC Software was developed with an applications focus for superior usability
- Resolution PC software is for use with all Biochrom UV/Vis Spectrophotometers
- Data acquisition using a Quick Read module (Lite only)
- Quick Read and Quick Scan for fast measurements with limited setup
- Bespoke method development for your unique laboratory requirements
- Save, print or export your analysis as an Excel™, Extended Metafile, HTML, pdf, rich text format, text, Word or XPS file types
Specifications
Software | |
All Biochrom UV/Vis Spectrophotometers |