missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Research Grade Fetal Bovine Serum, US Sourced
Research Grade Fetal Bovine Serum that is US sourced and triple 0.1 μm filtered.
Supplier: Fisherbrand™ SH3A1177402
Description
- US Origin is compliant with current good manufacturing practices (cGMP).
- For research use only.
Specifications
Gamma Globulin, Total Bilirubin, Sodium, Potassium, Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Osmolality (Ph. Eur. 2.2.35), Blood Urea Nitrogen, Creatinine, Percent Saturation, Alkaline Phosphatase, Glucose, IgG - Nephelometer, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine | |
United States | |
Bottle | |
100 mL | |
Sterile-filtered | |
Research | |
For Further Manufacturing Or Research Use. Not For Diagnostic Or Therapeutic Use. | |
< 35 mg/dL |
Fetal | |
Liquid | |
Fetal Bovine Serum | |
Sterile-filtered | |
Triple-filtered, 0.1 μm | |
Yes | |
No | |
No |