Learn More
Applied Biosystems™ DNA Dilution Buffer
DNA Dilution Buffer, a component of the ResDNASEQ Residual DNA Quantitation kits, is offered separately here for those kit users who need additional elution buffer.
Supplier: Applied Biosystems™ 4405587C
Description
DNA Dilution Buffer, a component of the ResDNASEQ Residual DNA Quantitation kits, is offered separately here for those kit users who need additional elution buffer.
As with other products in the Pharma Analytics Sample Preparation product line, DNA Dilution Buffer:
- Is completely supported by our global manufacturing and distribution centers
- Has a Certificate of Analysis available
- Has a shelf-life of up to 12 months
Specifications
7 mL | |
Store at 2°C to 8°C. | |
Liquid | |
Dilution Buffer |
For Research Use Only. Not for use in diagnostic procedures.