Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
Supplier:  Thermo Scientific™ 75005725
 View more versions of this product
Catalog No. 75-005-725
Only null left
Explore available promotions
Description
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
  • O-rings for inner and outer seals
  • ClickSeal lids for optimal biocontainment
  • Available for a variety of sizes and models
Specifications
Specifications
ClickSeal Biocontainment Lid
MicroClick 24 x 2 Rotor
Product Suggestions
Product Suggestions