Learn More
FEATHER Replaceable Blade Scissors No.145
FEATHER Replaceable Blade Scissors No.145 includes a handle that can be used for all three different blade tip types: sharp/sharp, sharp/blunt and blunt/blunt and 1 set of replacable blades. The replaceable blade design eliminates the cost and trouble of resharpening.
Supplier: FEATHER 145SHSH
Description
- Made in Japan
- Sharp/Sharp or Sharp/Blunt or Blunt/Blunt styles available
- Stainless steel handle and blade
Specifications
Scissors with Replacable Blades | |
48 mm | |
Autoclavable |
132 mm | |
Stainless Steel | |
Sharp/Sharp |