Learn More
Stemgent™ Alvetex Well Insert Starter Kit DFS Item
Plates, inserts and Petri dish/holder units are individually sterile blister packed
Supplier:  Stemgent™ AVPKIT2
Catalog No. AVPKIT2
Only null left
Explore available promotions
Includes:
  • Six 6-well inserts
  • Six 12-well inserts
  • One well insert holder in a deep Petri dish
Description
Description
Specifications
Specifications
6 of 6-well inserts, 6 of 12-well inserts and one of deep Petri dish for 3D cell culture
Insert Starter Kit
Product Suggestions
Product Suggestions