missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2-Amino-1-propanol 98.0+%, TCI America™
$162.52 - $480.00
Chemical Identifiers
CAS | 35320-23-1 |
---|---|
Molecular Formula | C3H9NO |
Molecular Weight (g/mol) | 75.111 |
MDL Number | MFCD00064413 |
InChI Key | BKMMTJMQCTUHRP-GSVOUGTGSA-N |
Synonym | d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol |
PubChem CID | 80308 |
IUPAC Name | (2R)-2-aminopropan-1-ol |
SMILES | CC(CO)N |
Chemical Identifiers
35320-23-1 | |
75.111 | |
BKMMTJMQCTUHRP-GSVOUGTGSA-N | |
80308 | |
CC(CO)N |
C3H9NO | |
MFCD00064413 | |
d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol | |
(2R)-2-aminopropan-1-ol |
Specifications
35320-23-1 | |
173°C | |
MFCD00064413 | |
2735 | |
BKMMTJMQCTUHRP-GSVOUGTGSA-N | |
(2R)-2-aminopropan-1-ol | |
80308 | |
≥98.0% (GC,T) | |
Liquid |
Yellow | |
C3H9NO | |
5 mL | |
d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol | |
CC(CO)N | |
75.111 | |
75.11 | |
Reagent | |
(R)-(-)-2-Amino-1-propanol |
Safety and Handling
EINECSNumber : (9)-0122
TSCA : No
Recommended Storage : Refrigerator