missing translation for 'onlineSavingsMsg'
Learn More

Contec™ QuickConnect™ Mop Head Frames

Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ QuickConnect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.

$280.06 - $280.06

Specifications

Color Code White
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
For Use With (Application) Pharmaceutical and medical device manufacturing facilities, or other similar critical or controlled environments
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Equipment) Length (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Length (Metric) Price Quantity  
19-176-941
View Documents
Contec™
2652K
VertiKlean MAX Mop Wipe System 29 cm
Each for $280.06
Only null left
 
Description

Description

  • Offer maximum flexibility for cleaning in critical environments.
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
Specifications

Specifications

White
Autoclavable
Pharmaceutical and medical device manufacturing facilities, or other similar critical or controlled environments
Stainless Steel
Non-sterile
Reusable
No
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'