Learn More
Qorpak™ HDPE F-Style Jugs — With Cap
With molded panel for easy label application
$119.32 - $269.81
Specifications
Color White
Includes Molded panel
Material HDPE
Closure Color White
Mouth Narrow
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
02-991-664
View Documents
Qorpak™
PLC03463
960 mL 24/Cs.
Case of 24 Each for $269.81
Only null left
02-991-705
View Documents
Qorpak™
PLC03461
3785 mL 6/Cs.
Case of 6 Each for $119.32
Only null left
Description
Description
  • Rectangular shape
  • Provide moisture barrier
  • Impact resistant
  • For high-speed filling, pharmaceutical uses and rough handling during shipping
  • Available in volumes ranging from 16 to 128 oz. (480 to 3785mL)
  • Jugs without cap also available (Cat. No. 02-991-167)
Specifications
Specifications
White
HDPE
Narrow
Polyethylene Foam
38-400 mm
Molded panel
White
F-style
Polypropylene
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'