missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Clear Glass Jugs without Cap

Storage solution for large quantity of liquids

$88.15 - $88.15

Specifications

Color Clear
Includes Molded finger grip
Material Glass
Mouth Narrow
Shape Cylindrical
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
02993012
View Documents
Qorpak™
GLA00838
3785 mL 4/Cs.
Case of 4 Each for $88.15
Only null left
 
Description

Description

  • Features molded finger grip
Specifications

Specifications

Clear
Glass
Cylindrical
Screw Cap
Molded finger grip
Narrow
38-400 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'