missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, Reagent, ACS, 99%, Spectrum™ Chemical
P1450, 110-86-1, C5H5N
Supplier: Spectrum Chemical Mfg Cor P1450500MLGLCS6
Description
Spectrum™ Chemical Pyridine, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
110-86-1 | |
6 x 500 mL | |
2e-005 | |
0.001 | |
C1=CC=NC=C1 | |
79.10 |
1 | |
Amber Glass Bottle | |
C5H5N | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
99% |
Chemical Identifiers
110-86-1 | |
79.10 | |
pyridine |
C5H5N | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
C1=CC=NC=C1 |