missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Reagent, ACS, 99%, Spectrum™ Chemical

P1450, 110-86-1, C5H5N

Supplier:  Spectrum Chemical Mfg Cor P145020LT

 View more versions of this product

Catalog No. 18605974

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Pyridine, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Specifications

Specifications

110-86-1
20 L
2e-005
0.001
C1=CC=NC=C1
79.10
ACS
1
Steel Can
C5H5N
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
99%

Chemical Identifiers

110-86-1
79.10
pyridine
C5H5N
JUJWROOIHBZHMG-UHFFFAOYSA-N
C1=CC=NC=C1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions