missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrobromide, 98%

CAS: 18820-82-1 | C5H6BrN | 160.014 g/mol

Supplier:  Thermo Scientific Chemicals A193140B

 View more versions of this product

Catalog No. AAA193140B


Only null left
Explore available promotions

Description

Description

Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications

Specifications

Pyridine hydrobromide
216°C to 220°C
MFCD00012801
Hygroscopic
Soluble in water.
C1=CC=NC=C1.Br
160.014
160.01
18820-82-1
C5H6BrN
1000 g
pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at
BBFCIBZLAVOLCF-UHFFFAOYSA-N
pyridine;hydrobromide
87810
98%

Chemical Identifiers

18820-82-1
160.014
BBFCIBZLAVOLCF-UHFFFAOYSA-N
87810
C1=CC=NC=C1.Br
C5H6BrN
MFCD00012801
pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at
pyridine;hydrobromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 242-600-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only