missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ PVC-Coated Wire Rack for 300mL BOD Bottles

This rugged wire rack holds up to (12) 300mL bottles for drying or storage.

Supplier:  DWK Life Sciences™ 227731

Catalog No. 36-102-5061


Only null left
Explore available promotions

Description

Description

  • Manufactured from rugged PVC coated wire
  • Coating is corrosion resistant
  • Racks interlock to connect stacked racks for safe transport
  • Made in the United States
Specifications

Specifications

Bottle Rack
Bottles (12 x 300 mL)
Yes, Interlocking
Case of 1
Corrosion-resistant rack
PVC-coated Wire
BOD bottles
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions