missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocol™ Zinc Formalin, Fisherbrand™
Ready-to-use buffered zinc formalin in bulk packaging.
$50.77 - $200.00
Specifications
For Use With (Equipment) | All Tissue Processors |
---|---|
Grade | Histological |
Product Type | Formalin |
Quantity | 1 gal. |
Solution Type | Formalin |
Form | Solution |
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
11-002-224
|
Fisherbrand
11002224 |
1 gal. |
Each for $50.77
Case of 4 Each for $203.08
|
|
|||||
11-002-225
|
Fisherbrand
11002225 |
5 gal. |
Each for $200.00
|
|
|||||
Description
- Especially recommended for use in immunohistochemistry, as zinc formalin preserves more immunoreactivity than standard NBF.
- Used as a fixative solution that provides excellent preservation of tissue morphology, particularly nuclear and cytoplasmic components, by increasing the rigidity of cellular structures
Specifications
All Tissue Processors | |
Formalin | |
Formalin |
Histological | |
1 gal. | |
Solution |
Made in the USA