missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Proteinase K, Tritirachium album
Serine protease that exhibits strong proteolytic activity on a wide variety of denatured and native proteins of high molecular weight
$85.10 - $187.00
Specifications
Formulation | Lyophilized from calcium acetate |
---|---|
Format | Lyophilized solid |
pH | Optimum 7.5 - 12.0 |
Content And Storage | Store at+2°C to+8°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
53948025MG
|
MilliporeSigma™
53948025MG |
25mg |
Each for $85.10
|
|
|||||
539480100MG
|
MilliporeSigma™
539480100MG |
100mg |
Each for $187.00
|
|
|||||
Description
- Because of its rapid proteolytic inactivation of endogenous nucleases, it can be used for isolation of mRNA and high molecular weight DNA
- Preferentially cleaves bonds next to the carbonyl group of N-substituted hydrophobic, aliphatic, and aromatic amino acids
- Inhibited by DFP, Hg2+, and PMSF
- Not inactivated by metal chelators, sulfhydryl reagents, TLCK, or TPCK
Specifications
Lyophilized from calcium acetate | |
Lyophilized solid | |
Optimum 7.5 - 12.0 | |
Store at+2°C to+8°C |