missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 10 Hooded Coveralls, Gray
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
$101.32 - $155.50
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | SMS Fabric |
Color | Gray |
Disposable | Single-use |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
18-888-712
|
DuPont™
PB127SGY2X002500 |
2X-Large |
Case of 25 Each for $107.69
|
|
|||||
18-888-713
|
DuPont™
PB127SGY3X002500 |
3X-Large |
Case of 25 Each for $114.08
|
|
|||||
18-888-714
|
DuPont™
PB127SGY4X002500 |
4X-Large |
Case of 25 Each for $123.63
|
|
|||||
18-888-715
|
DuPont™
PB127SGY5X002500 |
5X-Large |
Case of 25 Each for $133.18
|
|
|||||
18-888-717
|
DuPont™
PB127SGY7X002500 |
7X-Large |
Case of 25 Each for $155.50
|
|
|||||
18-888-718
|
DuPont™
PB127SGYLG002500 |
Large |
Case of 25 Each for $101.32
|
|
|||||
18-888-720
|
DuPont™
PB127SGYXL002500 |
X-Large |
Case of 25 Each for $101.32
|
|
|||||
18-888-716
|
DuPont™
PB127SGY6X002500 |
6X-Large |
Case of 25 Each for $144.00
|
|
|||||
Description
- Spunbond meltblown spunbond (SMS) coveralls have been optimized for comfort, softness, and breathability.
- Designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications.
- Applications include: general maintenance, janitorial/cleaning and other dirty work.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
- Attached hood with elastic around face opening.
- Elastic opening for tighter fit at wrist.
- Elastic opening for tighter fit at ankle.
- Manufactured under specifications that do not contain natural rubber latex.
Specifications
General Purpose Coveralls | |
SMS Fabric | |
Single-use | |
Elastic | |
Attached | |
Serged | |
Particle | |
PB127SWH | |
General Maintenance, Janitorial, Cleaning |
Unisex | |
Gray | |
Elastic | |
Hooded | |
Front Zipper | |
Non-sterile | |
Yes | |
None | |
25/Cs. |