missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Phenoxytol, Reagent, Spectrum™ Chemical
PR132, 770-35-4, C9H12O2
Supplier: Spectrum Chemical Mfg Cor PR13220LT
Description
Spectrum™ Chemical Propylene Phenoxytol, ReagentSpecifications
770-35-4 | |
242 to 244°C | |
C9H12O2 | |
20 L | |
CC(O)COC1=CC=CC=C1 | |
152.19 | |
Reagent |
1 | |
Poly Bottle | |
MFCD00016861 | |
IBLKWZIFZMJLFL-UHFFFAOYNA-N | |
1-phenoxypropan-2-ol | |
93% | |
Liquid |
Chemical Identifiers
770-35-4 | |
152.19 | |
IBLKWZIFZMJLFL-UHFFFAOYNA-N | |
CC(O)COC1=CC=CC=C1 |
C9H12O2 | |
MFCD00016861 | |
1-phenoxypropan-2-ol |