missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol, USP, 99.5%, Spectrum™ Chemical
PR130, 57-55-6, C3H8O2
Supplier: Spectrum Chemical Mfg Cor PR1304LTPLCS4
Description
Spectrum™ Chemical Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
0.20 mL | |
1 | |
Poly Bottle | |
4 x 4 L | |
CC(O)CO | |
76.10 |
57-55-6 | |
3.5 mg/50g | |
C3H8O2 | |
DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
propane-1,2-diol | |
99.5% |
Chemical Identifiers
57-55-6 | |
76.10 | |
propane-1,2-diol |
C3H8O2 | |
DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
CC(O)CO |