missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Carbonate, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P1433200LTBL
Specifications
108-32-7 | |
Drum | |
0.99 |
1 | |
200 L | |
Reagent |
Chemical Identifiers
108-32-7 |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.