missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Propidium Iodide, Calbiochem™,
$287.00 - $645.00
Chemical Identifiers
CAS | 25535-16-4 |
---|---|
Molecular Formula | C27H34I2N4 |
Molecular Weight (g/mol) | 668.406 |
InChI Key | XJMOSONTPMZWPB-UHFFFAOYSA-M |
Synonym | propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide |
PubChem CID | 104981 |
ChEBI | CHEBI:51240 |
IUPAC Name | 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide |
SMILES | CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
537059100MG
|
MilliporeSigma™
537059100MG |
100 mg | Glass Bottle |
Each for $287.00
|
|
||||
537059250MG
|
MilliporeSigma™
537059250MG |
250 mg | Glass Bottle |
Each for $645.00
|
|
||||
Chemical Identifiers
25535-16-4 | |
668.406 | |
propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide | |
CHEBI:51240 | |
CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-] |
C27H34I2N4 | |
XJMOSONTPMZWPB-UHFFFAOYSA-M | |
104981 | |
3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide |
Specifications
25535-16-4 | |
Glass Bottle | |
100 mg | |
XJMOSONTPMZWPB-UHFFFAOYSA-M | |
3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide | |
104981 | |
Solid |
Red | |
C27H34I2N4 | |
propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide | |
CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-] | |
668.406 | |
CHEBI:51240 |