missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 900 Half-Finger Impact Gloves

AVC™ padding in the palm and other key areas helps to reduce shock and impact. Coating on the palm provides enhanced gripping capabilities.

$74.41 - $74.41

Specifications

Color Black
Hand Specific
Material Spandex
Product Type Anti-impact Gloves
For Use With (Application) Construction, Manufacturing, Hand and Power Tools
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-820-198
View Documents
Ergodyne™
17692
Small
Pair for $74.41
Only null left
 
19-820-199
View Documents
Ergodyne™
17693
Medium
Pair for $74.41
Only null left
 
19-820-200
View Documents
Ergodyne™
17694
Large
Pair for $74.41
Only null left
 
19-820-201
View Documents
Ergodyne™
17695
X-Large
Pair for $74.41
Only null left
 
19-820-202
View Documents
Ergodyne™
17696
2X-Large
Pair for $74.41
Only null left
 
Description

Description

  • Segmented AVC™ palm padding reduces shock and palm impact
  • Gripping palm provides secure hold on tools and equipment
  • Half-finger design for dexterity and tactile feedback
  • Hex stretch knit and breathable spandex construction
  • Neoprene knuckle pad with a reinforced thumb saddle
  • Low-profile molded hook & loop closure
  • Gloves include ID space to write user's names
  • Finger webbing loops for easy on/off
Specifications

Specifications

Black
Spandex
Construction, Manufacturing, Hand and Power Tools
ProFlex™ 900
Specific
Anti-impact Gloves
Hook and Loop
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'