missing translation for 'onlineSavingsMsg'
Learn More

Proflavine Sulfate, Hydrate, Spectrum™ Chemical

$515.80 - $1533.00

Chemical Identifiers

CAS 1811-28-5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612949
View Documents
Spectrum Chemical Mfg Cor
P142925GM
25 g Amber Glass Bottle
Each for $515.80
Only null left
 
18612950
View Documents
Spectrum Chemical Mfg Cor
P1429100GM
100 g Amber Glass Bottle
Each for $1,533.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1811-28-5

Specifications

1811-28-5
Amber Glass Bottle
Reagent
1
25 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.