Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human BTLA (CD272) Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Supplier: Invitrogen™ EPX01A12217901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to human BTLA (CD272)
- human BTLA (CD272) Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human BTLA (CD272) monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
96 Tests | |
Multiplex Immunoassay | |
Cell Culture Supernatant, Plasma, Serum | |
B and T lymphocyte attenuator |
Human | |
CD272 (BTLA) | |
Serum, Plasma: 25 μL; CCS: 50 μL |