missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PrepSEQ™ Rapid Spin Sample Preparation Kit
Supplier: Thermo Fisher Scientific 4407760
Specifications
This kit includes 100 columns, 100 1.5 mL tubes (store at 22° to 28°C), and Lysis buffer (store at 2° to 8°C). These reagents are sufficient for 100 samples. | |
Coarse Filtration, Spin Column | |
Bacterial DNA | |
Real-Time Quantitative PCR (qPCR), PCR | |
100 | |
Rapid Spin Sample Preparation Kit | |
Genomic DNA |
Spin Column | |
Swab, Bacteria (Gram -), Bacterial Culture, Food, Water | |
Centrifuge | |
Not High-throughput Compatible (Manual) | |
PrepSEQ™ | |
1 Kit | |
750 μL |