Learn More
ABS Premium Laboratory Freezer
Delivering superior cooling to laboratory environments.
Supplier:  ABS ABTHCPLF23
 View more versions of this product
Catalog No. 361058608
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Engineered with variable speed compressors (VSCs), these units feature ultra-quiet operation and significant energy savings. VSCs optimize energy consumption by self-adjusting to cooling demands. These compressors also deliver enhanced system performance and provide a longer lifespan than other compressor variations.
  • Ultra-quiet variable speed compressors
  • Superior energy efficiency
  • Microprocessor temperature control
  • Full array of alarms
  • Natural hydrocarbon refrigerants
Enjoy the uniformity and speed of microprocessor temperature control and a full array of alarms to safeguard your products. Unit controllers also come with a battery backup to keep your data safe. Upgrade your laboratory environment with these premium freezers and welcome energy savings, noise reduction, smooth operation, and improved system performance to your work environment.
Specifications
Specifications
Laboratory Freezer
651 L
Solid Swing
Scratch-resistant Painted Steel
5
Upright
NEMA 5-15
Variable Speed
23 cu. ft.
-35°C to -15°C
1
Stainless Steel
ETL
Alarm Module, Battery Back-up
115 V
Product Suggestions
Product Suggestions