missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Certified, 1.00N ±0.01N (1.0M), LabChem™

$131.43 - $756.53

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC198151
View Documents
LabChem
LC198151
500 mL
Each for $131.43
Only null left
 
LC198152
View Documents
LabChem
LC198152
1 L
Each for $231.53
Only null left
 
LC198154
View Documents
LabChem
LC198154
4 L
Each for $756.53
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7681-11-0
166.003
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide

Specifications

7681-11-0,7732-18-5
85.15,14.85
500 mL
KI
Soluble in water
[K+].[I-]
166.003
CHEBI:8346
Certified
Passes Test
1.11g/mL
1.11g/mL
Yellow
Liquid
IK
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166
1.00N ±0.01N (1.0M)
Amber Poly
Iodine vapor
Potassium Iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes eye irritation.

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature; Protect from light

SDS
missing translation for 'documents'

missing translation for 'documents'