missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Pellets, NF, Spectrum™ Chemical
PO180, 1310-58-3, KOH
$472.77 - $4782.17
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
IUPAC Name | potassium hydroxide |
SMILES | [OH-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18605601
|
Spectrum Chemical Mfg Cor
PO180500GM |
500 g | Poly Bottle |
Each for $472.77
|
|
||||
18605603
|
Spectrum Chemical Mfg Cor
PO18025KG |
2.5 kg | Poly Bottle |
Each for $1,521.90
|
|
||||
18605602
|
Spectrum Chemical Mfg Cor
PO180500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,355.10
|
|
||||
18605604
|
Spectrum Chemical Mfg Cor
PO18025KGCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $4,782.17
|
|
||||
18605605
|
Spectrum Chemical Mfg Cor
PO18012KG |
12 kg | Poly Bottle |
Each for $3,199.27
|
|
||||
18605606
|
Spectrum Chemical Mfg Cor
PO18050KGBL |
50 kg | Poly Bottle |
Each for $4,353.03
|
|
||||
Description
Spectrum™ Chemical Potassium Hydroxide, Pellets, NF is used as a pH modifier. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
[OH-].[K+] |
HKO | |
MFCD00003553 | |
potassium hydroxide |
Specifications
1310-58-3 | |
HKO | |
500 g | |
[OH-].[K+] | |
56.11 | |
NF | |
Potassium Hydroxide |
100% | |
MFCD00003553 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
85% | |
Poly Bottle |