missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
$36.30 - $443.70
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium hydroxide |
SMILES | [OH-].[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
LC193001
|
LabChem
LC193001 |
500 mL |
Each for $36.30
|
|
|||||
LC193002
|
LabChem
LC193002 |
1 L |
Each for $49.87
|
|
|||||
LC193004
|
LabChem
LC193004 |
4 L |
Each for $145.27
|
|
|||||
LC193006
|
LabChem
LC193006 |
10 L |
Each for $297.00
|
|
|||||
LC193005
|
LabChem
LC193005 |
20 L |
Each for $443.70
|
|
|||||
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Specifications
Colorless | |
99.4,0.6 | |
KOH | |
0.1000N ±0.0005N (0.1M) | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
500 mL | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Certified | |
Poly Bottle | |
Traceable to NIST | |
Potassium Hydroxide |
1310-58-3,7732-18-5 | |
HKO | |
MFCD00003553 | |
1g/mL | |
Soluble in water | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
Passes Test | |
1g/mL | |
Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature