missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 45% (w/w), Analytical Reagent Grade, Ricca Chemical
$213.59 - $2737.50
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium hydroxide |
SMILES | [OH-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6190-16
|
Ricca Chemical Company
619016 |
500 mL | Natural Poly Bottle |
Each for $213.59
|
|
||||
6190-32
|
Ricca Chemical Company
619032 |
1 L | Natural Poly Bottle |
Each for $307.63
|
|
||||
6190-5
|
Ricca Chemical Company
61905 |
20 L | Cubitainer |
Each for $2,737.50
|
|
||||
R619000055E
|
Ricca Chemical Company
R619000055E |
55 gal. | Poly Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Specifications
Colorless | |
98.0 | |
HKO | |
45% (w/w) | |
500 mL | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Analytical | |
Aqueous Solution | |
Potassium Hydroxide |
1310-58-3,7732-18-5 | |
100.0 | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
Min. 45.0% | |
Natural Poly Bottle | |
Liquid |
Safety and Handling
Recommended Storage : Corrosive