missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.500 N (N/2), Ricca Chemical
$129.63 - $1148.25
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium hydroxide |
SMILES | [OH-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
6230-16
|
Ricca Chemical Company
623016 |
500 mL | Natural Poly Bottle |
Each for $129.63
|
|
||||
6230-32
|
Ricca Chemical Company
623032 |
1 L | Natural Poly Bottle |
Each for $182.47
|
|
||||
6230-1
|
Ricca Chemical Company
62301 |
4 L | Natural Poly Bottle |
Each for $441.19
|
|
||||
6230-5
|
Ricca Chemical Company
62305 |
20 L | Cubitainer |
Each for $1,148.25
|
|
||||
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Specifications
Colorless | |
95.0,3.06 | |
HKO | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
500 mL | |
0.500N |
1310-58-3,7732-18-5 | |
99.0,3.19 | |
MFCD00003553 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
Natural Poly Bottle | |
Liquid | |
Potassium Hydroxide |