missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.100 N (N/10) in Methanol, Ricca Chemical
$168.41 - $578.97
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium hydroxide |
SMILES | [OH-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
626016
|
Ricca Chemical Company
626016 |
500 mL | Natural Poly Bottle |
Each for $168.41
|
|
||||
6260-32
|
Ricca Chemical Company
626032 |
1 L | Natural Poly Bottle |
Each for $213.09
|
|
||||
6260-1
|
Ricca Chemical Company
RI62601 |
4 L | Natural Poly Bottle |
Each for $578.97
|
|
||||
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Specifications
Colorless | |
97.0,0.8 | |
HKO | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Methanol | |
Liquid | |
Potassium Hydroxide |
1310-58-3,67-56-1 | |
100.0,0.83 | |
MFCD00003553 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
Natural Poly Bottle | |
500 mL | |
0.100N |