missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogen Phthalate, 100mg/L, Reagents
Potassium Hydrogen Phthalate, 100mg/L, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$200.23 - $406.27
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Potassium Acid Phthalate, KHP, Potassium Biphthalate |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-334-259
|
Labchem Inc
CP1931001C |
1 L |
Each for $200.23
|
|
|||||
01-334-258
|
Labchem Inc
CP1931004C |
4 L |
Each for $406.27
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Potassium Acid Phthalate, KHP, Potassium Biphthalate | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
4 | |
100°C | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
Glass Bottle | |
1 |
7732-18-5,877-24-7 | |
Colorless | |
Potassium Acid Phthalate, KHP, Potassium Biphthalate | |
O | |
18.02 | |
1 L | |
Liquid | |
Potassium Hydrogen Phthalate, 100 mg/L |