missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferrocyanide, Trihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$22318.89 - $22318.89
Chemical Identifiers
CAS | 14459-95-1 |
---|---|
Molecular Weight (g/mol) | 422.39 |
MDL Number | MFCD00167023 |
InChI Key | NLYKFUFIUWPPIZ-UHFFFAOYSA-N |
IUPAC Name | tetrapotassium hexacyanoironbis(ylium) trihydrate |
SMILES | O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
02003878
|
Avantor J.T.Baker
311405 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
14459-95-1 | |
MFCD00167023 | |
tetrapotassium hexacyanoironbis(ylium) trihydrate |
422.39 | |
NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
Specifications
14459-95-1 | |
2.5 kg | |
MFCD00167023 | |
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N | |
422.39 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 1.85kg |
Crystals | |
C6H6FeK4N6O3 | |
NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
tetrapotassium hexacyanoironbis(ylium) trihydrate | |
422.41 | |
Poly Bottle | |
Potassium Ferrocyanide, Trihydrate |