Learn More
Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals
$439.56 - $439.56
Chemical Identifiers
CAS | 888711-44-2 |
---|---|
Molecular Formula | CH2BBrF3K |
Molecular Weight (g/mol) | 200.835 |
MDL Number | MFCD09265154 |
InChI Key | AZDFPIRYUOCVCJ-UHFFFAOYSA-N |
Synonym | potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v |
PubChem CID | 23690312 |
IUPAC Name | potassium;bromomethyl(trifluoro)boranuide |
SMILES | [B-](CBr)(F)(F)F.[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3216806
|
Thermo Scientific Chemicals
H3216806 |
5 g |
Each for $439.56
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
888711-44-2 | |
200.835 | |
AZDFPIRYUOCVCJ-UHFFFAOYSA-N | |
23690312 | |
[B-](CBr)(F)(F)F.[K+] |
CH2BBrF3K | |
MFCD09265154 | |
potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v | |
potassium;bromomethyl(trifluoro)boranuide |
Specifications
888711-44-2 | |
BrCH2BF3K | |
5 g | |
AZDFPIRYUOCVCJ-UHFFFAOYSA-N | |
potassium;bromomethyl(trifluoro)boranuide | |
23690312 | |
95% |
CH2BBrF3K | |
MFCD09265154 | |
potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v | |
[B-](CBr)(F)(F)F.[K+] | |
200.835 | |
200.84 | |
Potassium bromomethyltrifluoroborate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only