missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bisulfate, Puriss. p.a., ≥99%, Honeywell Fluka™
Puriss. p.a.,≥99%
$750.60 - $750.60
Chemical Identifiers
CAS | 7646-93-7 |
---|---|
Molecular Formula | HKO4S |
Molecular Weight (g/mol) | 136.16 |
MDL Number | MFCD00011404 |
InChI Key | CHKVPAROMQMJNQ-UHFFFAOYSA-M |
Synonym | potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c |
PubChem CID | 516920 |
IUPAC Name | potassium;hydrogen sulfate |
SMILES | [K+].OS([O-])(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7646-93-7 | |
136.16 | |
CHKVPAROMQMJNQ-UHFFFAOYSA-M | |
516920 | |
[K+].OS([O-])(=O)=O |
HKO4S | |
MFCD00011404 | |
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c | |
potassium;hydrogen sulfate |
Specifications
7646-93-7 | |
HKO4S | |
MFCD00011404 | |
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c | |
[K+].OS([O-])(=O)=O | |
136.16 | |
136.17g/mol | |
puriss. p.a. | |
Potassium Bisulfate |
1 kg | |
KHSO4 | |
2509 | |
CHKVPAROMQMJNQ-UHFFFAOYSA-M | |
potassium;hydrogen sulfate | |
516920 | |
≥99% | |
2.32g/mL at 25°C |
Safety and Handling
P261-P280-P305 + P351 + P338-P310
H314-H335
EINECSNumber : 231-594-1
RTECSNumber : TS7200000