missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bis(fluorosulfonyl)imide 95.0+%, TCI America™
$139.84 - $411.97
Chemical Identifiers
CAS | 14984-76-0 |
---|---|
Molecular Formula | F2KNO4S2 |
Molecular Weight (g/mol) | 219.218 |
MDL Number | MFCD17018950 |
InChI Key | MHEBVKPOSBNNAC-UHFFFAOYSA-N |
PubChem CID | 23697682 |
IUPAC Name | potassium;bis(fluorosulfonyl)azanide |
SMILES | [N-](S(=O)(=O)F)S(=O)(=O)F.[K+] |
Chemical Identifiers
14984-76-0 | |
219.218 | |
MHEBVKPOSBNNAC-UHFFFAOYSA-N | |
potassium;bis(fluorosulfonyl)azanide |
F2KNO4S2 | |
MFCD17018950 | |
23697682 | |
[N-](S(=O)(=O)F)S(=O)(=O)F.[K+] |
Specifications
14984-76-0 | |
White | |
MFCD17018950 | |
1759 | |
[N-](S(=O)(=O)F)S(=O)(=O)F.[K+] | |
219.218 | |
219.22 | |
Crystalline Powder |
101°C | |
F2KNO4S2 | |
5 g | |
MHEBVKPOSBNNAC-UHFFFAOYSA-N | |
potassium;bis(fluorosulfonyl)azanide | |
23697682 | |
≥95.0% (T) | |
Potassium Bis(fluorosulfonyl)imide |
Safety and Handling
TSCA : No