missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene 2-place Slide Mailer

Store or ship two standard slides with confidence. Polypropylene 2-Place Slide Mailer has side-by-side slots in a sturdy polypropylene shell. Large, easy to secure snaps provide secure storage. Disposable mailers are available in a variety of colors for easy coding.

Catalog No. 14-127-51



Only null left
Explore available promotions

Description

Description

  • Protectors hold two standard slides: 1 x 3 in. or 25 x 75 mm
  • Large, easy snap lids ensure slides stay securely in the mailer
  • Slides lay flat in separate channels to avoid slide contact or shifting
  • Variety of colors for easy coding
  • 25 disposable mailers per pack
Specifications

Specifications

68 mm
2.7 in.
Standard Microscope Slides
0.2 in.
5 mm
2 Slides
3.7 in.
93 mm
Assorted
Slide Mailer
3.7 x 2.7 x 0.2 in (93 x 68 x 5 mm)
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions