missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ Polypropylene Coveralls with Elastic Wrists and Open Ankles
Disposable coveralls provide low-cost protection against dust, dirt, and grime.
Supplier: International Enviroguard™ 20124XL
Description
- Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
- Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
- Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
- Elastic wrists, open ankles
- Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
- Stay cool, comfortable, and protected from head-to-toe
- Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Light-duty Coveralls | |
Unisex | |
White | |
Standard | |
Elastic | |
Unhooded | |
Serged | |
Particle, Dust | |
Yes | |
None |
4X-Large | |
Polypropylene | |
Single-use | |
Open | |
Collared | |
Front Zipper | |
Non-sterile | |
Fold-down | |
2012 | |
Food Packaging, Food Processing, Warehousing, Laboratories |