missing translation for 'onlineSavingsMsg'
Learn More

Polygalacturonic acid, >86%, MP Biomedicals™

$520.86 - $520.86

Chemical Identifiers

CAS 25990-10-7
Molecular Formula (C6H8O6)n
Molecular Weight (g/mol) 194.14
MDL Number MFCD00131972
InChI Key AEMOLEFTQBMNLQ-BKBMJHBISA-N
Synonym galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate
PubChem CID 445929
ChEBI CHEBI:33885
IUPAC Name (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10271125
View Documents
MP Biomedicals Inc
0210271125
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

25990-10-7
194.14
AEMOLEFTQBMNLQ-BKBMJHBISA-N
445929
(2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid
(C6H8O6)n
MFCD00131972
galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate
CHEBI:33885
O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O

Specifications

25990-10-7
>86%
MFCD00131972
galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate
O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O
194.14
CHEBI:33885
88.7%
Polygalacturonic acid, from Pectin
Tan to White
(C6H8O6)n
25 g
AEMOLEFTQBMNLQ-BKBMJHBISA-N
(2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid
445929
(176)n n=150 to 400
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature (15 to 30°C).

SDS
missing translation for 'documents'

missing translation for 'documents'