missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 3350, USP, 97-103%, Spectrum™ Chemical

PO125, 25322-68-3, H(OCH2CH2)nOH

$923.33 - $923.33

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605320
View Documents
Spectrum Chemical Mfg Cor
PO12525KG
2.5 kg Poly Pail N/A
Only null left
 
Description

Description

Spectrum™ Chemical Polyethylene Glycol 3350, USP acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
[H]OCCO
(C2H4O)n
MFCD01779601
ethane-1,2-diol

Specifications

25322-68-3
(C2H4O)n
2.5 kg
[H]OCCO
62.07
USP
0.001
0.01
1
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
97 to 103%
30-38
Poly Pail
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'