missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polyester Sign: KEEP ELECTRICAL PANEL CLEAR FOR 36 INCHES
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$19.19 - $19.19
Specifications
Material | Polyester |
---|---|
Shape | Rectangular |
Displayed Text | KEEP ELECTRICAL PANEL CLEAR FOR 36 INCHES |
Color | Yellow on White |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
19-109-602
|
Brady™
144433 |
7 x 10 in. |
Each for $19.19
|
|
|||||
19-109-603
|
Brady™
144434 |
10 x 14 in. |
Each for $19.19
|
|
|||||
Description
- Polyester material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Polyester | |
Rectangular | |
KEEP ELECTRICAL PANEL CLEAR FOR 36 INCHES | |
Yellow on White |